Nunc™ Polyolefin Acrylate Sealing Tape, Thermo Scientific
Catalog #: CA37000-548
Supplier: Thermo Fisher Scientific
Specifications
- Color:Clear
- Description:Amplification Tape
- Cat. no.:CA37000-548
- Supplier No.:232702
Specifications
About this item
The polyolefin acrylate sealing tapes minimize evaporation and protect samples from contamination and spilling.
- Use for PCR, fluorescence assays, and microscopy applications
- Pressure sensitive adhesive
- Non-pierceable
- Nonsterile
Ideal for all assay applications.
Certifications: Certified RNase- and DNase-free.
Ordering information: Compatiable with 96- and 384-well plates.
Customers Who Bought This Also Bought