Order Entry
Canada
ContactUsLinkComponent
Borate Buffered Saline Milk Blocking Solution-10X, BioFX®, Surmodics IVD, Inc.
Borate Buffered Saline Milk Blocking Solution-10X, BioFX®, Surmodics IVD, Inc.
Catalog # 101396-992
Supplier:  Surmodics
CAS Number:  
Borate Buffered Saline Milk Blocking Solution-10X, BioFX®, Surmodics IVD, Inc.
Catalog # 101396-992
Supplier:  Surmodics
Supplier Number:  MLKB-1000-01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Borate Buffered Saline Milk Blocking Solution-10X
  • Size:
    1 L
  • Environmentally Preferable:
  • Protein:
    Bovine
  • Shelf life:
    3 years
  • Storage buffer:
    Borate buffered saline
  • Storage temperature:
    2…8 °C
  • Cat. no.:
    101396-992
  • Supplier No.:
    MLKB-1000-01

Specifications

About this item

BioFX® Borate Buffered Saline (BBS) Milk Blocking Solution-10X Concentrate contains Non-Fat Dry Milk (NFDM) in a BBS solution that is supplied as a solution ready for dilution to specific application requirements.

  • 3 year shelf life
  • Multiple applications
  • Ready to use formulation

This product serves as a blocking reagent when diluted in reagent quality water. As a blocking reagent, the solution binds to the secondary protein binding sites following the binding of the primary antigen or antibody to the solid phase surface.

Certifications: ISO 13485:2016 & 9001:2015 Certified