If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:STEM Investigations: Fingerprint Classification
- No. of Lab Groups:15
- Cat. no.:470311-678
- Includes:Teacher's Manual and Student Guide
Real world STEM investigation.
- Additional optional activities included
- Students create classification system
In this lab activity students will learn about the history of fingerprinting as a tool for identification. They then will analyze fingerprints for common characteristics and create a classification system using those characteristics.
Once they have created a system, they will analyze class data using the system that is used worldwide for classification. This kit includes an optional activity where students can practice lifting a fingerprint and comparing it to a 'crime scene' print. Kit contains enough materials for 15 groups. Teacher’s guide and student guide copymasters are included.