ContactUsLinkComponent
Reagent Reservoirs, 12 Column, Integra Biosciences
Reagent Reservoirs, 12 Column, Integra Biosciences
Catalog # MSPP-6361
Supplier:  INTEGRA MS
Reagent Reservoirs, 12 Column, Integra Biosciences
Catalog # MSPP-6361
Supplier:  INTEGRA MS
Supplier Number:  6361
Specifications
  • Capacity:
    21 ml
  • Description:
    12 Column Reagent Reservoirs
  • Cat. no.:
    MSPP-6361
  • Supplier No.:
    6361
Specifications
About this item
The polypropylene design improves the reservoirs chemical compatibility and also allows them to be autoclaved.
  • Reagent reservoirs for 96 well plates
  • Pyramid bottom
  • Each column holds 21 ml