Sample Vials, Clear, Without Caps, Kimble Chase, DWK Life Sciences
About this item
Screw thread sample vials are manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Class A requirements.
Ordering information: Closures are not included.
Packaging: Vials are packaged in corrugated trays with partitions.
12 Options Available Below
Product Details & Documents
Screw thread sample vials are manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Class A requirements.
Ordering information: Closures are not included.
Packaging: Vials are packaged in corrugated trays with partitions.
Product Family Options
Product Information
- CapacityDiameter ×HMaterialCap includedCap SizeAvailabilityPrice
- Supplier Number: 60812-1232
- Supplier Number: 60812-1235
- Supplier Number: 60812-1528
- Supplier Number: 60812-1545
- Supplier Number: 60812-1738
- Supplier Number: 60812-1760
- Supplier Number: 60812-1940
- Supplier Number: 60812-1965
- Supplier Number: 60812-2170
- Supplier Number: 60812-2385
- Supplier Number: 60812-2870
- Supplier Number: 60812-2895