$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Racks
Tube Racks
Storage Rack for Vials, Polyethylene, Chemglass
Storage Rack for Vials, Polyethylene, Chemglass
75820-618
:
Chemglass
Storage Rack for Vials, Polyethylene, Chemglass
75820-618
:
Chemglass
:
CV-6005-0012
No. of Holes:
50
Material:
Polyethylene
Environmentally Preferable:
Cat. no.:
75820-618
For Tubes:
12×32 mm
Supplier No.:
CV-6005-0012
:
Vials and closures must be purchased separately.