Closures, Polypropylene, Chemglass
Supplier: Chemglass
Certificates
About this item
Closure offers two positions, open for gas exchange or closed for humidified environment.
- Internal drip ring minimizes contamination
- Autoclavable to 121 °C
Customers Who Bought This Also Bought