Order Entry
Canada
ContactUsLinkComponent
Betatrophin (139-198) (Human) - EIA Kit, Phoenix Pharmaceuticals
Betatrophin (139-198) (Human) - EIA Kit, Phoenix Pharmaceuticals
  MSPP-EK05155
 :  
Betatrophin (139-198) (Human) - EIA Kit, Phoenix Pharmaceuticals
  MSPP-EK05155
 :  EK-051-55
 :  

 

  • Assay Type:
    Competitive
  • Primary antibody reactivity:
    Human
  • Target protein:
    Betatrophin (139-198)
  • Description:
    Betatrophin (139-198) (Human)
  • Size:
    96 wells
  • Storage temperature:
    4 °C
  • Sensitivity:
    0.28 ng/ml
  • Cat. no.:
    MSPP-EK05155
  • Supplier No.:
    EK-051-55

 

 

This kit is designed to measure the concentration of Betatrophin (139-198) and its related proteins based on the principle of a competitive enzyme immunoassay.

 : The kit contains EIA assay buffer concentrate (20X, 50 ml), pre-coated EIA plate (96 wells, 1 plate), acetate plate sealer (APS) (3 pieces), primary betatrophin (139-198) antibody (1 vial), standard betatrophin (139-198) (1 vial), biotinylated betatrophin (139-198) (1 vial), positive control (2 vials), streptavidin-horseradish peroxidase (SA-HRP) concentrate (30 µl), substrate solution (TMB) (12 ml), 2 N HCl (stop solution) (15 ml), protocol booklet (1 book).