Order Entry
Canada
ContactUsLinkComponent
VWR® Carboy with Closed Cap, Clear PC
VWR® Carboy with Closed Cap, Clear PC
  76199-180
VWR® Carboy with Closed Cap, Clear PC
  76199-180

 

  • Capacity:
    10 L
  • Height:
    19.7"
  • Length:
    10.75"
  • Cat. no.:
    76199-180
  • Weight:
    6.38"

 

 

The ergonomically body is coupled with a VersaCap® technology to provide the user an easy-to-use and easy-to-clean carboy.

  • Easy-to-read metric graduation marks certified to ±5% accuracy
  • Molded-in material identification markings

Carboy’s body includes molded hand grips in the base to allow the user to easily hold and manipulate during lifting, filling, and pouring.

The polycarbonate carboy is made from a clear, autoclavable, USP Class VI, FDA Grade resin. It is suitable for aqueous solutions, as well as most life sciences buffer solutions.

The product also offers 120 mm neck to simplify filling and cleaning. They are compatible with the VersaCap® technology for interchangeable top connections.