Order Entry
ContactUsLinkComponent
Dropette® Disposable Transfer Pipettes, Simport Scientific
Dropette® Disposable Transfer Pipettes, Simport Scientific
Catalog #: CA11027-120
Supplier:  Simport Scientific
Dropette® Disposable Transfer Pipettes, Simport Scientific
Catalog #: CA11027-120
Supplier:  Simport Scientific

Specifications

  • Capacity:
    3 ml
  • Material:
    Polyethylene
  • Cat. no.:
    CA11027-120
  • Overall Length:
    13.8 cm
  • Bulb Draw:
    2.1 ml
  • Supplier No.:
    P200-30

Specifications

About this item

These all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.

  • Can be sealed and refrigerated
  • Temperature resistant down to –196 °C
  • Can be gas (EtO) sterilized
  • Non-toxic and inert
  • No bulb to insert or remove
  • Uniform drop size

Put an end to matching rubber bulbs with glass pipettes. Molded from see-through low density polyethylene. Inert to biological fluids and most acids. The low-affinity surface reduces the loss of cells and valuable proteins due to binding.

Customers Who Bought This Also Bought