Graduated Transfer Pipettes, Bio Nuclear Diagnostics
Catalog #: CA53300-905
Supplier: Bio Nuclear Diagnostics
Specifications
- Capacity:7.7 ml
- Material:Polyethylene
- Cat. no.:CA53300-905
- Graduations:3 ml
- Bulb Draw:3.2 ml
- Supplier No.:PIP-005
Specifications
About this item
General-purpose transfer pipettes have a built-in bulb that eliminates the need for plugging, packaging, and autoclaving, which prevents cross-contamination. Graduation markings allow measuring of more than one volume.
- Made of shatterproof, non-toxic and inert low density polyethylene
- Low-affinity surface reduces loss of cells and valuable proteins due to binding
- Can be frozen in liquid nitrogen, or heat sealed for storage
Recommended for use in tissue culture applications, routing sampling of serum and plasma, blood culture, and urine analysis.
Customers Who Bought This Also Bought