Measuring (Mohr) Pipettes with Colored Markings, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
Mohr pipettes are calibrated to class A specifications and USP standards for volumetric glassware. Pipettes are designed to deliver total capacity without blow-out.
- Easily identify size by color-coded graduations enameled into the glass
- Top end of 5 to 25 ml sizes is constricted (Mohr type)
- Reusable
Frequently Bought Together