Absorbent Pad Petri Dishes, Simport®
Supplier: Simport Scientific
Certificates
About this item
These 50×9 mm Petri dishes are ideal for culturing micro-organisms on either agar or broth based media.
- Petri dish with absorbent pad
- Accomodates 47 mm diameter membrane filter
- Made of polystyrene
- Stackable
- Sterile
Designed to accommodate 47 mm diameter membrane filters, these polystyrene dishes are stackable and feature squared-off edges on both the top and bottom to provide convenient grips for one-handed opening. The snug fitting top ensures that neither the absorbent pad nor the agar media will dry out during incubation. A frosted area on top of the lid permits labeling.
Packaging: Dishes are packaged in heavy-wall polyethylene sleeves.
Caution: Not for tissue culture application.
Customers Who Bought This Also Bought