Buffer Assortment Pack, Hydrion Buffer Chemvelopes, Micro Essential Lab
Product Details & Documents
Buffer assortment pack includes buffers with pH ranges from 2 to 11. Chemvelopes are made of a triple protective foil consisting of aluminum, polyethylene and paper which keeps the Hydrion Buffers dry and free-flowing.
Hydrion buffer chemvelopes produce 500 ml of pH buffer standard solution when dissolved in distilled water.
All buffers are also available separately.
Specifications
Specifications
- Pack type:Pack
- Description:Buffer Assortment Pack
- pH:2 - 11
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



