Orders LinkContactUsLinkComponent
Filtration Assembly
Filtration Assembly
Filtration Assembly
Catalog # 470310-690
Supplier:   Wards
Supplier #:  FSAS16
Filtration Assembly
Catalog # 470310-690
Supplier:  Wards
Filtration Assembly
Filtration Assembly
Filtration Assembly
Price
$439.80
Log in for availability
Each
Product Details & Documents
This filtration assembly is used for vacuum filtration of tissue culture media, biological fluids, HPLC and other aqueous solutions.
  • Unit is designed to maximize flow and reduce foaming
  • Versatile and effective
Assembly consists of 300 ml borosilicate glass funnel, highly integrated sealed 47 mm fritted glass support base, one litre flask. An aluminum clamp is provided to hold the filter funnel and support base properly. The 47 mm filter provides an effective filtration area of 9.6 square centimeters.
Specifications
  • Description:
    Filtration Assembly