Order Entry
Canada
ContactUsLinkComponent
Dissecting Forceps, Serrated Tips, Walter Stern
Dissecting Forceps, Serrated Tips, Walter Stern
Catalog # 25607-195
Supplier:  Walter Stern
Dissecting Forceps, Serrated Tips, Walter Stern
Catalog # 25607-195
Supplier:  Walter Stern
Supplier Number:  300-130

Specifications

  • Length:
    114 mm (4.5")
  • Material:
    Stainless steel
  • Cat. no.:
    25607-195
  • Supplier No.:
    300-130

Specifications

About this item

Stainless steel dissecting forceps. Straight body, with fine points, serrated tips and guide pins.