Orders LinkContactUsLinkComponent
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
These vials are used with Beckman™, CTC™, Gilson™, Knauer™, Shimadzu™, Spark™, Varian™ and Hitachi™ autosamplers that require a narrow neck dimensions.
  • Type-1, N51A Expansion borosilicate glass
  • High quality, low cost with reliable performance
Useful in GC or HPLC.
Product Family Options
  • Description
    Capacity
    Color
    Cap included
    Cap Size
  • Screw Top Vials
    2.00 ml
    Amber
    No
    8-425
    Pack of 100
    Pack of 100 - $57.11
    $57.11
    Pack of 100
       $57.11
     : 9582S-0AV
  • Screw Top Vials with Write On Patch
    2.00 ml
    Amber
    No
    8-425
    Pack of 100
    Pack of 100 - $75.85
    $75.85
    Pack of 100
       $75.85
     : 9582S-WAV
  • Screw Top Vials
    2.00 ml
    Clear
    No
    8-425
    Pack of 100
    Pack of 100 - $49.08
    $49.08
    Pack of 100
       $49.08
     : 9582S-0CV
  • Screw Top Vials with Write On Patch
    2.00 ml
    Clear
    No
    8-425
    Pack of 100
    Pack of 100 - $65.85
    $65.85
    Pack of 100
       $65.85
     : 9582S-WCV
  • Screw Top Vials, Tear Drop
    1.10 ml
    Clear
    No
    8-425
    Pack of 100
    Pack of 100 - $210.17
    $210.17
    Pack of 100
       $210.17
     : 9582S-11-0CV
Inserts
  •  
    Color
    Bottom style
    Diameter ×H
    Cap included
    Capacity
    Description
  • Clear
    Flat bottom
    5×31 mm
    No
    0.20 ml
    Inserts
    Case of 1,000
    Case of 1,000 - $619.60
    $619.60
    Case of 1,000
       $619.60
  •  
    Color
    Bottom style
    Diameter ×H
    Cap included
    Capacity
    Description
  • Clear
    Conical bottom
    5×29 mm
    No
    0.10 ml
    Inserts with attached plastic spring
    Pack of 100
    Pack of 100 - $218.84
    $218.84
    Pack of 100
       $218.84