Eyewear Cleaning Fluid, HexArmor
76207-854
: HexArmor
- Description:Eyewear cleaning fluid
- Size:8 oz.
- Cat. no.:76207-854
Eyewear cleaning fluid, chemically formulated to ensure TruShield® coatings continuously perform to the highest standard.
- 8 oz. cleaning fluid
- Suitable for all lenses
- Fits into wall mount cleaning station
Proprietary coatings are chemically formulated to perform differently than any other coating on the market. Therefore, using standard, non-HexArmor, alcohol-based cleaner on them can cause damage.
For this reason, specific cleaning supplies are available to ensure the TruShield® coatings continuously perform to the highest standard. Using any anti-fog wipes, alcohol-based cleaners, or any other non-HexArmor product on TruShield® lenses can cause damage.