Orders LinkContactUsLinkComponent
Ward's® Mini Magnifier
Ward's® Mini Magnifier
Mini magnifier, 16.1×13.5×20.7 cm
Catalog # 470311-198
Supplier:   Heathrow Scientific
Supplier #:  HS15092D
Ward's® Mini Magnifier
Catalog # 470311-198
Ward's® Mini Magnifier
Ward's® Mini Magnifier
Mini magnifier, 16.1×13.5×20.7 cm
Price
215,70 $
Log in for availability
Each Case of 24
Product Details & Documents
Mini magnifier enlarges objects for easier view.
  • Swiveling, molded lens provides 1.75× magnification
  • Excellent for viewing and counting colonies on bacterial plates
The Ward's® mini magnifier is mounted on a stand to keep hands free for manipulating samples. Excellent for viewing bacterial colonies on media, the magnifier pairs well with the Ward's® mini light box and Ward's® benchtop counter lines (both available separately).
Specifications
  • Magnification:
    1.75×
  • Description:
    Mini magnifier, 16.1×13.5×20.7 cm