TLS Preset Torque Screwdriver, Mountz
Catalog # 76247-738
Supplier: MOUNTZ INC TE
Specifications
- Description:Female Hex Drive Model, Minor
- Cat. no.:76247-738
- Handle Color:Red
- Torque:1.4 - 13.7 kgf.cm (1.2 - 12" ft.lbs.)
- Drive Size:1/4"
Specifications
About this item
TLS is a preset torque limiting screwdriver (internally adjustable). Built with a cam-over design, the accurate torque screwdriver prevents a fastener or bolt from being over-torqued.
- Precision radial ball clutch slips-free when the preset torque is reached preventing over-torquing
- Thrust bearings insure that the torque setting is independent of any end load applied by the operator
- Tamper-proof internal adjustment. No external adjustment scale - must be preset using a torque analyzer
- Provide bi-directional operation and are calibrated in either the CW or CCW direction
When the torque screwdriver reaches the pre-set torque value the internal mechanism disengages from the drive thus limiting the torque applied.