Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Tool Sets
Morse® Cutting Tool, Cutting Tool Supply
Morse® Cutting Tool, Cutting Tool Supply
76253-726
:
CUTTING TOOL SUPPLY TE
Morse® Cutting Tool, Cutting Tool Supply
76253-726
:
CUTTING TOOL SUPPLY TE
:
91157
Description:
Cutting Tool, 4 Flute
Length:
2½"
Material:
Carbide
Size:
Finishing
Cat. no.:
76253-726
Type:
Single End
Finish:
ALTiN
Shank Diameter:
¼"
Diameter:
1
/
4
"