ContactUsLinkComponent
Ground Cord for Floor Mat, Menda
Ground Cord for Floor Mat, Menda
Catalog # 76242-576
Supplier:  Menda
undefined
Ground Cord for Floor Mat, Menda
Catalog # 76242-576
Supplier:  Menda
Supplier Number:  09816
Specifications
  • Color:
    Green/Yellow
  • Description:
    Banana Plug Ground Cord,10 mm Snap Stud
  • Cat. no.:
    76242-576
Specifications
About this item
Ground cord terminated with a banana plug, easily snaps to mat socket, flush to mat surface.
Stud has threaded hole; 6 to 32×1/4" flat head screw and countersunk washer are provided allows ground cord to be optionally bolted mat; keeps cord from disconnecting; ensures proper grounding.
Certifications: Meets grounding standard ANSI/ESD S6.1.