End Cap Clamp, Holland Applied Technologies
Catalog # 76237-894
Supplier: Holland Applied Technologies
Specifications
- Description:End Cap Clamp
- Material:Stainless steel
- Size:3/4"
- Cat. no.:76237-894
Specifications
About this item
Solid end cap is made of stainless steel 304 and clamps on to the end of a process line. The end cap clamps on to the end of a ferruled tube to close the line, providing a non-permanent seal that provides take down access.
This sanitary clamp end cap meets 3-A sanitary standards and is suitable for use in food, beverage, dairy and cosmetic applications.