Orders LinkContactUsLinkComponent
HY-PRO® NRT® Taps, Sid Tool Co
HY-PRO® NRT® Taps, 0.141" Ø shank, 9/16" thread size, 0.110" (square), 1⅞"
Catalog # 76253-800
Supplier:   SID TOOL CO INC TE
 :  9247875
HY-PRO® NRT® Taps, Sid Tool Co
Catalog # 76253-800
Supplier:  SID TOOL CO INC TE
HY-PRO® NRT® Taps, Sid Tool Co
HY-PRO® NRT® Taps, 0.141" Ø shank, 9/16" thread size, 0.110" (square), 1⅞"
Price
$141.61
Log in for availability
Each
Product Details & Documents
Forming tap with UNC thread.
  • Tin coated
  • Thread limit H2
  • Threads 40 per inch
  • Chamfer 2.5 P
Specifications
  • Description:
    HY-PRO® NRT® Taps, 0.141" Ø shank, 9/16" thread size
  • Length:
    1⅞"
  • Size:
    0.110" (square)