QuadraTest Multimode Electronic Test Indicator, Sid Tool
Electronic Test Indicator
Catalog # 76253-182
Supplier: SID TOOL CO INC TE
Supplier #: 20426136
QuadraTest Multimode Electronic Test Indicator, Sid Tool
Catalog # 76253-182
Supplier: SID TOOL CO INC TE
Jump to:
Product Details & Documents
Multimode electronic test indicator offers unrivaled measuring versatility in an electronic test indicator!
Available with a 1/2" or 17/16" contact point. The QuadraTest is the perfect solution to test indicating applications.
Features four measuring modes (Minmum, Maximum, TIR and direct), with an RS-232 interface that is water, oil and coolant resistant to IP65 compliancy with or without the interface cable installed! The QuadraTest also features true inch/metric conversion, a 11/2" diameter dial face that rotates a full 360° with both analog bar graph and numeric digital display.
Certifications: Meets or exceeds DIN/ISO standards.
Specifications
- Description:Electronic Test Indicator
- Cat. No.:76253-182
- Measuring range:1/5"/0.5 mm