ContactUsLinkComponent
Camß Digital Inspection Magnifier, Vision Engineering
Camß Digital Inspection Magnifier, Vision Engineering
Catalog # 76252-774
Camß Digital Inspection Magnifier, Vision Engineering
Catalog # 76252-774
Supplier Number:  CMB001
Specifications
  • Magnification:
    4×, 8×, 16×, 20×
  • Description:
    Cam ß Digital Inspection Magnifier
  • Cat. no.:
    76252-774
Specifications
About this item
This is designed especially for industrial applications where viewing, image capture, reporting and documentation is needed. Camβ is optimized for use in a secure environments where Bluetooth or communication capable devices are not acceptable. The Camβ builds on the substantial success of its predecessor, the CamZ.
  • Highly versatile and portable design
  • Large 4.3" color display
  • Simple to operate
  • Store up to 20000 photos
  • Live view via HDMI output
Optics and illumination optimized for close subjects.