ContactUsLinkComponent
Grommet Snap Installation Hand Tool, Desco Industries
Grommet Snap Installation Hand Tool, Desco Industries
Catalog # 76246-452
Grommet Snap Installation Hand Tool, Desco Industries
Catalog # 76246-452
Supplier Number:  PRES-N-SNAP TOOL
Specifications
  • Description:
    PRES-N-Snap Tool
  • Cat. no.:
    76246-452
Specifications
About this item
This is a versatile tool used to install grommet style snaps.
  • Heat-treated cast aluminum construction
  • Lightweight and durable allows for one-handed operation
Capable of setting numerous brands of fasteners simply by changing the dies.