Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stoppers
PE/LDPE/HDPE Stoppers
Connector Plug Stereo, Digi-Key
Connector Plug Stereo, Digi-Key
Connector Plug Stereo, Digi-Key
Catalog # 76251-440
Supplier:
DIGI-KEY CORPORATION
Expand image
Connector Plug Stereo, Digi-Key
Catalog # 76251-440
Supplier:
DIGI-KEY CORPORATION
Supplier Number:
TT253Z
Specifications
Color:
Gold and Black
Description:
Connector Phone Plug Stereo
Height:
4.39 mm
Cat. no.:
76251-440
Specifications
About this item
Product Details
Product Details