Slide Switch, Digi-Key
Catalog # 76250-376
Supplier: DIGI-KEY CORPORATION
Specifications
- Voltage:125 V
- Color:Silver
- Material:Copper
- Cat. no.:76250-376
- Operating Temperature:–10...104 °C
Specifications
About this item
Miniature panel mount and through hole slide switch.
- Vertical orientation
- Two through five positions
- Detent or spring return