Eclipse Punchdown Tool, Global Industrial
Eclipse 110 Punchdown Tool, Black/blue
Catalog # 76248-634
Supplier: GLOBAL INDUSTRIAL TE
Supplier #: CP-3145A
Jump to:
Product Details & Documents
ABS grip handle and light weight design provides user comfort.
- Perfect for small jobs and the do-it-yourself installer
Rotatable blade holder for easy selection of proper blade type end.
Specifications
- Color:Black/blue
- Description:Eclipse 110 Punchdown Tool