Acetone 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Catalog # CAJT2462-03
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Poly bottle, pressure bottom
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 solvent selection guide recommended after discussion
- Cat. no.:CAJT2462-03
Specifications
Specification Test Results
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Specific Gravity at 25°/25°C:≤0.789
- NF - Nonvolatile Residue:≤0.004 %
- NF - Readily Oxidizable Substances:Passes Test
- NF - Water (H₂O):≤0.5 %
- NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis):99.5 - 100.5 %
- NF - Benzene:≤2 ppm
- ACS - Aldehydes (as HCHO):≤0.002 %
- ACS - Assay (CH₃COCH₃) (by GC, corrected for water):≥99.5 %
- ACS - Color (APHA):≤10
- ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤0.05 %
- ACS - Methanol (CH₃OH)(by GC):≤0.05 %
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Titrable Base (meq/g):≤0.0006
- ACS - Water (H₂O):≤0.5 %
Specification Test Results
About this item
:
:58.08 g/mol
UN:1090
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765