ContactUsLinkComponent
PureFlo® MKP Series Polyethylene Filter Capsules, Zenpure Americas
PureFlo® MKP Series Polyethylene Filter Capsules, Zenpure Americas
Catalog # 76250-034
PureFlo® MKP Series Polyethylene Filter Capsules, Zenpure Americas
Catalog # 76250-034
Supplier Number:  MKPS020B2H2H
Specifications
  • Pore size:
    0.2 µm
  • Description:
    PureFlo® MKP Series Polyethylene Filter Capsules
  • Material:
    PE
  • Cat. no.:
    76250-034
Specifications
About this item
PureFlo® MKP Series Polyethylene (PE) capsules were designed for small to medium-scale filtration, clarification, and venting applications in the pharmaceutical, biotechnology, food and beverage, medical and water industries worldwide. This family of products is particularly suitable for venting applications where the units need to be gamma sterilized with other disposable units for venting. The hydrophobic PE membrane is ideally suited for air, gas and liquid sterilizing applications.
The hydrophobic membrane provides superior flow and pressure-drop characteristics for small tank and bio-bag venting applications. The MKP capsules have a compact design that can incorporate 400 cm² of filtration material to meet the needs of a wide variety of processes. We are currently offering four different polyethylene media sizes with 24 different fitting options. The units are thermally-sealed. No adhesives or binders are used in the encapsulation process. The units are rinsed with pyrogen-free water to reduce extractables and downtime. All filter capsules are 100% integrity-tested to ensure filter performance each and every time out of the package.