PALL Filter Element Replacement, Filtermart
Catalogue # 76267-644
Fournisseur: FILTERMART TE
Spécifications
- Description:PALL Filter Element Replacement
- Cat. no.:76267-644
Spécifications
À propos de cet article
Particulate filters, also known as after-filters, are used to remove solid particulate contamination from compressed air line.
They are typically located after a compressed air desiccant dryer. These compressed air particulate filters provide the highest level of clean compressed air with a minimum loss of pressure. Compressed air particulate filter elements are tough enough to hold their shape under pressure and maintain even pressure differentials to avoid collapsing of the filter element.