ContactUsLinkComponent
Sanitary Tri-Clamp® Standard Gaskets, Garlock Hygienic Technologies
Sanitary Tri-Clamp® Standard Gaskets, Garlock Hygienic Technologies
Catalog # 76238-142
Supplier:  Rubber Fab
Sanitary Tri-Clamp® Standard Gaskets, Garlock Hygienic Technologies
Catalog # 76238-142
Supplier:  Rubber Fab
Supplier Number:  40MPU-150
Specifications
  • Color:
    Black
  • Material:
    Buna
  • Size:
    1 - 1/2"
  • Cat. no.:
    76238-142
Specifications
About this item
Sanitary gaskets are used widely in the food, dairy, beverage, biotech, pharmaceutical, and all other sanitary process industries to seal clamp connections in sanitary pipe lines.
Rubber fab sanitary Tri-Clamp® gaskets are ADI free (Animal Derived Ingredient free). Buna N is not USP class VI or ADI free.