Aquasyn Diaphragms, Holland Applied Technologies
76249-792
- Description:Diaphragm, silicone, 1/4" through 1/2" valve size
- Cat. no.:76249-792
Aquasyn's diaphragms are precision molded in the open position to tight tolerances for superior service life and made of approved materials which will not affect the purity of the most sensitive of processes.
EPDM diaphragms suitable for the temperature range –70 to 130 °C.
: USP and FDA compliant.