Orders LinkContactUsLinkComponent
Chamfer Tools, Cutting Tool Supply
Chamfer Tools, Cutting Tool Supply
MCH-2R Single End and MCH-2D Double End Chamfer Tool, with 2 Flutes, Steel, 3", ½", 82°, ½"
Catalog # 76249-864
Supplier #:  B10226
Chamfer Tools, Cutting Tool Supply
Catalog # 76249-864
Chamfer Tools, Cutting Tool Supply
MCH-2R Single End and MCH-2D Double End Chamfer Tool, with 2 Flutes, Steel, 3", ½", 82°, ½"
Price
$202.00
Log in for availability
Each
Product Details & Documents
Chamfer tools made of solid carbide.
Suitable for cutting steel, cast iron and non-ferrous materials.
Specifications
  • Description:
    MCH-2R Single End and MCH-2D Double End Chamfer Tool, with 2 Flutes
  • Length:
    3"
  • Material:
    Steel
  • Shank diameter:
    ½"
  • Cutting diameter:
    ½"
  • Ratcheting angle:
    82°