Klein® Cable Cutter, TESTEQUITY LLC TE
Catalog # 76250-926
Supplier: TESTEQUITY LLC TE
Specifications
- Description:Cable Cutter
- Cat. no.:76250-926
- Overall Length:9½"
- Cut Type:Shear cut
Specifications
About this item
High leverage design and shear-type jaws cut up to 4/0 aluminum, up to 2/0 soft copper, and up to 100 pair, 24 AWG communication cable.