ContactUsLinkComponent
Crimp Tool with Y501 Die Set, DMC
Crimp Tool with Y501 Die Set, DMC
Catalog # 76253-222
Supplier:  DMC TE
Crimp Tool with Y501 Die Set, DMC
Catalog # 76253-222
Supplier:  DMC TE
Supplier Number:  HX4-501
Specifications
  • Description:
    Crimp Tool with Y501 Die Set
  • Cat. no.:
    76253-222
Specifications
About this item
This open frame crimp tool is comes with pre-installed Y501 die set.
  • For use with pre-insulated terminal crimp 26-10 AWG