ContactUsLinkComponent
Astro Hand Crimp Tool, Newark/Element
Astro Hand Crimp Tool, Newark/Element
Catalogue # 76277-120
Fournisseur:  NEWARK/ELEMENT 14 TE
Astro Hand Crimp Tool, Newark/Element
Catalogue # 76277-120
Fournisseur:  NEWARK/ELEMENT 14 TE
Supplier Number:  M22520/7-01
Spécifications
  • Description:
    Hand Crimp Tool
  • Cat. no.:
    76277-120
Spécifications
À propos de cet article
This crimp tool is specially designed to use with MIL-DTL-38999 series III contacts.