Molex Ratchet Crimp Tool, Newark/Element
Catalog # 76253-836
Supplier: NEWARK/ELEMENT 14 TE
Specifications
- Description:Ratchet Crimp Tool
- Cat. no.:76253-836
Specifications
About this item
This full cycle ratcheting hand tool ensures complete crimps and has ergonomic soft grip handles for comfortable crimping.
- Right and left-handed applications
- For Mini-Fit™ Jr. male and female crimp terminals, 22 to 28AWG
A precision user-friendly terminal locator wire stop holds terminals in the proper crimping position.
Certifications: Tool is IPC/WHMA A-620 Class 2 compliant.