WHEATON® Headspace Crimp Top Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
WHEATON® Headspace Crimp Top Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
Jump to:
About this item
Headspace vials are manufactured from clear Type I borosilicate glass.
- Accept 20 mm aluminum seals
- Flat bottom and beveled finish
Ordering information: Caps not included (sold separately).
2 Options Available Below
Product Details & Documents
Headspace vials are manufactured from clear Type I borosilicate glass.
- Accept 20 mm aluminum seals
- Flat bottom and beveled finish
Ordering information: Caps not included (sold separately).
Documents
Product Family Options
Product Information
- CapacityDiameter ×HMaterialColorCap includedAvailabilityPrice
- 89495-248Vials, Beveled FinishVials, Beveled Finish6.00 ml22×38 mmBorosilicate glassClearNoLog in for availabilityCase of 100$77.02Supplier Number: W225281
- 89495-250Vials, Beveled FinishVials, Beveled Finish10.00 ml23×46 mmBorosilicate glassClearNoDiscontinued-Supplier Number: -
- 89495-252Vials, Beveled FinishVials, Beveled Finish20.00 ml23×75 mmBorosilicate glassClearNoLog in for availabilityCase of 100$99.45Supplier Number: W225283