Gas Detector Tubes for Ethyl Ether, Sensidyne
Catalog # 13502-608
Supplier: Sensidyne
Small Business Enterprise
Specifications
- Temperature Range:0...40 °C
- Cat. no.:13502-608
- Measurement Range:0.04 - 1.4%
- No. of Strokes:1 (100 ml)
- Color Change:Orange to dark green
- Detectable Gas:Ethyl ether
- Supplier No.:107SA
Specifications
About this item
Colorimetric gas detector tubes are used for detection of airbourne gases and vapors.
- Contains an antibacterial grip
- Easy to use
These detector tubes safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors at a very low cost-per sample. These tubes are easy to use and provide a low-cost method of routine or spot sampling for gases and vapors.