Gas Detector Tubes for Ethyl Alcohol, Sensidyne
Catalog #: 14231-012
Supplier: Sensidyne
Small Business Enterprise
Specifications
- Measurement range (°Brix):0.05−5.0%
- Measurement range:0.05−5.0%
- Temperature Range:0...40 ºC
- No. of Strokes:1 (100 ml)
- Color Change:Yellowish orange to pale green
- Detectable Gas:Ethyl alcohol
- Supplier No.:104SA
Specifications
About this item
Gas detector tubes are used for detection of airbourne gases and vapors.
- Contains an antibacterial grip
- Easy to use
These detector tubes safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors at a very low cost-per sample. These tubes are easy to use and provide a low-cost method of routine or spot sampling for gases and vapors.
Customers Who Bought This Also Bought