ContactUsLinkComponent
Permanent ESD Safe Foams, Conductive Containers
Permanent ESD Safe Foams, Conductive Containers
Catalog # 76246-066
Permanent ESD Safe Foams, Conductive Containers
Catalog # 76246-066
Supplier Number:  CFM3939-25
Specifications
  • Color:
    Black
  • Size:
    39×39×1/4"
  • Cat. no.:
    76246-066
Specifications
About this item
Static safe foams in sheet form have permanent ESD safe properties.
Permanent black foams are ideal for shelf liners, tote cushions, long-term storage, or reusable shipping applications.