
Barbituric acid ≥98%, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
CAS Number:67-52-7
:
Formula:C₄H₄N₂O₃
MDL Number:MFCD00006666
:128.09 g/mol
Merck Index:13,00966
Boiling Pt:441 °C (1013 hPa)
Melting Pt:250…252 °C
Density:0.95 g/cm³
:Management of Change (MOC) category = N
EINECS:200-658-0
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Loss on Drying at 105°C:≤2 %
- Solubility (4% in H₂O):Passes Test
- Assay (C₄H₄N₂O₃) (by acid-base titration):≥98 %
- Appearance:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot NumberB654 Product Specifications
Additional Product Details
sds_BAKRB654
SDS
Frequently Bought Together