Break-over Wrenches with Fixed Heads, Mountz
Supplier: MOUNTZ INC TE
Certificates
About this item
These wrenches are designed and manufactured to meet or exceed the accuracy and repeatability requirements of ISO 6789:2003 (6% of setting).
- Small and lightweight
- Tamper-proof internal adjustment
Breaks-over when preset torque is reached eliminating overtorquing. The break-over mechanism minimizes shock to assemblies. No external adjustment scale must be preset using a torque analyzer. When the TB wrench achieves torque, the head breaks-over preventing the operator from overtorquing the fastener.
Frequently Bought Together