Ideal® Wire Stripper, T®-5, TESTEQUITY LLC TE
76243-438
- Description:Ergonomic wire stripper
- Cat. no.:76243-438
- Overall Length:7"
T-5 Wire stripper for 10 to 18 AWG size (solid) and 12 to 20 (stranded) AWG size wires.
T-5 Wire stripper for 10 to 18 AWG size (solid) and 12 to 20 (stranded) AWG size wires.