Ground Cord, Male Stud, SCS, Desco Industries
Catalog # 89024-538
Supplier: Desco
Specifications
- Description:Common ground cord, 1 meg resistor
- Length:15'
- Cat. no.:89024-538
- Closure:10 mm, male
- Supplier No.:CGC151M
Specifications
About this item
This ground cord connects two wrist straps and a worksurface to ground.
- Compatible with mats that have a 10 mm socket snap
- The 10 ring terminal at end of ground cord allows easy attachment to electrical ground
- No resistor is required, but with resistor is acceptable
Delivery: Includes 1 megohm resistor, meets Grounding standard ANSI/ESD S6.1.
Frequently Bought Together