ContactUsLinkComponent
6 Point Deep Impact SAE Sockets, Apex
6 Point Deep Impact SAE Sockets, Apex
Catalog # 76250-084
Supplier:  Cooper Tools
6 Point Deep Impact SAE Sockets, Apex
Catalog # 76250-084
Supplier:  Cooper Tools
Supplier Number:  80373
Specifications
  • Description:
    Deep SAE Socket, 7/8"
  • Cat. no.:
    76250-084
  • No. of Points:
    6
Specifications
About this item
Chrome molybdenum alloy steel (Cr-Mo) for exceptional strength and durability.
  • Off-corner loading design reduces fastener rounding
  • Outer coating to resist corrosion
Certifications: Meets or Exceeds ASME B107.1.