To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:IgG1
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.5 mg
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below Avoid cycles of freezing and thawing. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:immunoaffinity chromatography
- Cat. no.:76226-086
Specifications
About this item
Camelids produce functional IgG isotypes that do not incorporate light chains. Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. At present, three subclasses of camelid IgG have been identified (IgG1, IgG2, IgG3), of which IgG2 and IgG3 lack the light chains. IgG2 consists of dimers of short heavy chains, which are characterized by a normal Fc region without a CH1 domain. IgG2 shows a molecular weight of approximately 90 kDa and this smaller size improves its bio-distribution and allows a better tissue penetration. Of special importance is the long complementary determining region (CDR) loop which inserts deep into the active site of an enzyme enabling a complete neutralization of an enzyme. Anti-Llama IgG2 generated in rabbit detects specifically Llama IgG2 isotype. This anti-Llama secondary Antibody is suitable for western blot, ELISA, ChIP and immunohistochemistry as well as other more general immunoassays.
Antibody Anti-Llama IgG2 is suitable for immunoblotting (western or dot blot), ELISA, and immunohistochemistry assays requiring lot-to-lot consistency.
Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Camel